Learn More
American Research Products Inc Mouse Thyroid PeroXidase TPO
Catalog No. 5000329441
Encompass
Mouse Thyroid-PeroXidase TPO ELISA Kit from CUSABIO Detection Range 2 5 ng/mL-40 ng/mL Sensitivity 2 5 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-29441 Supplier American Research Products Inc Supplier No. CSBE08353M
Mouse Thyroid-PeroXidase TPO ELISA Kit from CUSABIO Detection Range 2 5 ng/mL-40 ng/mL Sensitivity 2 5 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive