Learn More
BIOHIPPO Mouse TICAM1 Microsample
SDP
Catalog No. 5000313774
Encompass
Mouse TICAM1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TICAM1 in tissue homogenates cell lysates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 23 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TICAM1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TICAM1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TICAM1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13774 Supplier BIOHIPPO Supplier No. BHE15210868
Mouse TICAM1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TICAM1 in tissue homogenates cell lysates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 23 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TICAM1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TICAM1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TICAM1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color