Learn More
Raybiotech Inc Mouse TIM-1 (KIM-1) ELISA Kit
Supplier:  Raybiotech Inc ELMTIM11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 30 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse B7-1, BAFF R, BTC, C5a, CCL6, CD48, CD6, Chemerin, Clusterin, CXCL15, Cystatin C, DAN, DLL4, EDAR, Endocan, Fetuin A, H60, IL-33, IL-7 R alpha, Kremen-1, Limitin, Lipocalin-2, LOX-1, Marapsin, MBL-2, Meteorin, Nope, NOV, Osteoactivin, OX40, Ligand, P-Cadherin, Periostin, PlGF-2, Progranulin, Prostasin, Renin 1, Testican 3, TIM-2, TIM-3, TRAIL, Tryptase epsilon
Catalog No. 50-195-0139