Learn More
Bon Opus Biosciences Recombinant Mouse Tim-2 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP103550UG
Encompass_Preferred
Recombinant Mouse Tim-2 (C-hFc) is produced by Human Cells. The target gene encoding His22-Asn229 is expressed with a C-hFc tag. Accession# NP_001154827.1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1035
Catalog No. 50-217-2634