Learn More
Bon Opus Biosciences Recombinant Mouse Tim-2 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP103650UG
Encompass_Preferred
Recombinant Mouse Tim-2 (C-mFc) is produced by Human Cells. The target gene encoding His22-Asn229 is expressed with a C-mFc tag. Accession# NP_001154827.1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1036
Catalog No. 50-217-2635