Learn More
Sino Biological Recombinant Mouse Tim4/TIMD4 Protein (His Tag) 1
Supplier:  Sino Biological 50388M08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse TIMD4 (NP_848874.3) precursor (Met 1-Thr 279) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-224-6947