Learn More
Sino Biological Anti-Tim4/TIMD4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50388RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TIMD4 (rM TIMD4; Catalog#50388-M08H; NP_848874.3; Met 1-Thr 279). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3290