Learn More
Sino Biological Recombinant Anti-TIMP-1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50342R008100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TIMP-1 (rM TIMP-1; Catalog#50342-MNAH; NP_035723.2; Met1-Arg205).
Catalog No. 50-255-3303