Learn More
Sino Biological Mouse TIMP1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 106164T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse TIMP-1, and purified by antigen affinity chromatography.
Catalog No. 50-237-1708