Learn More
Biomatik Corporation Mouse Tissue Factor Pathway Inhibitor 2 (TFPI2) ELISA Kit
Supplier:  Biomatik Corporation EKU11473
Encompass_Preferred
Mouse Tissue Factor Pathway Inhibitor 2 (TFPI2) ELISA Kit for detecting TFPI2 in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35536. Gene ID: 21789. Target Synonyms: PP5; Placental protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5602