Learn More
Biomatik Corporation Mouse Tissue Factor (TF) CLIA Kit
Supplier:  Biomatik Corporation EKU08875
Encompass_Preferred
Mouse Tissue Factor (TF) CLIA Kit for detecting TF in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.95pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P20352. Gene ID: 14066. Target Synonyms: CD142; FIII; F3; TFA; Thromboplastin; Coagulation Factor III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8608