Learn More
American Research Products Inc Mouse tissue inhibitors of met
Catalog No. 5000326784
Encompass
Mouse tissue inhibitors of metalloproteinase 3 TIMP-3 ELISA kit from CUSABIO Detection Range 46 88 pg/mL-3000 pg/mL Sensitivity 11 72 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26784 Supplier American Research Products Inc Supplier No. CSBE07385M
Mouse tissue inhibitors of metalloproteinase 3 TIMP-3 ELISA kit from CUSABIO Detection Range 46 88 pg/mL-3000 pg/mL Sensitivity 11 72 pg/mL Sample serum plasma tissue homogenates Method Sandwich