Learn More
Biomatik Corporation Mouse Tissue Inhibitors Of Metalloproteinase 4 (TIMP4) ELISA Kit
Supplier:  Biomatik Corporation EKU07732
Encompass_Preferred
Mouse Tissue Inhibitors Of Metalloproteinase 4 (TIMP4) ELISA Kit for detecting TIMP4 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.12-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JHB3. Gene ID: 110595. Target Synonyms: TIMP Metallopeptidase Inhibitor 4; Tissue Inhibitor of Metalloproteinases 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8730