Learn More
Abcam Mouse Tissue kallikrein ELISA Kit, 1 x 96 Tests
Catalog No. 502663560
Encompass_Preferred
Mouse Tissue kallikrein ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Tissue kallikrein with a sensitivity of 4.2 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-3560 Supplier Abcam Supplier No. AB2336301X96TESTS
Add to Cart
Add to Cart
Mouse Tissue kallikrein ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Tissue kallikrein with a sensitivity of 4.2 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement