Learn More
ACROBiosystems Mouse TL1A Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems TLAM52431MG
Encompass_Preferred
Mouse TL1A , His Tag (TLA-M5243) is expressed from human 293 cells (HEK293). It contains AA Ala 61 - Leu 252 (Accession # Q5UBV8-1).
Catalog No. 50-233-1988