Learn More
BIOHIPPO Mouse TLN Microsample
SDP
Catalog No. 5000315101
Encompass
Mouse TLN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TLN in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 07 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TLN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TLN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TLN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15101 Supplier BIOHIPPO Supplier No. BHE15211302
Mouse TLN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TLN in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 07 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TLN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TLN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TLN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color