Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse TLR11 Synthetic Peptide

Catalog No. PEP0176
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0176 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0176 Supplier Invitrogen™ Supplier No. PEP0176
Only null left
Add to Cart
Add to Cart

Synthetic Peptide

TLR11 Synthetic Peptide, PEP-0176, from Invitrogen.

The Toll-like receptor (TLR) family in mammal comprises a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and IL-1 receptor motif in the cytoplasmic domain. Like its counterparts in Drosophila, TLRs signal through adaptor molecules (1). The TLR family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition (2). TLR11 does not respond to known TLR ligands but instead respond specifically to uropathogenic bacteria. TLR11-deficient mice are highly susceptible to infection of the kidneys by uropathogenic bacteria, indicating that TLR11 may play a protective role in preventing infection of internal organs of the urogenital system.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 239081
Species Mouse
Purification Method purified
Accession Number Q6R5P0
Common Name Mouse TLR11
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol Tlr11
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.