Learn More
Invivogen Anti-mouse/human TLR2 mAb, clone T2.5, 100 ug
SDP
Supplier:  Invivogen MAB2MTLR2
Encompass
MAb mTLR2 (T2.5) is a monoclonal antibody that reacts with mouse Toll-like receptor 2 (TLR2, CD282). MAb mTLR2 is an antagonistic antibody. It has been tested by flow cytometric analysis of RAW264.7 cells. The antibody is cross reactive with human TLR2.
Catalog No. NC1378277