Learn More
Invivogen MOUSE TLR9 AGONIST KIT
SDP
Catalog No. NC1399059
Encompass
This kit contains archetypal CpG ODNs of the A-, B- or C-class described for mice cells immunostimulation via TLR9.
Catalog No. NC1399059 Supplier Invivogen Supplier No. TLRLKIT9M
This kit contains archetypal CpG ODNs of the A-, B- or C-class described for mice cells immunostimulation via TLR9.