Learn More
BIOHIPPO Mouse TM Microsample
SDP
Catalog No. 5000314215
Encompass
Mouse TM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TM in plasma It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14215 Supplier BIOHIPPO Supplier No. BHE15212603
Mouse TM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TM in plasma It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color