Learn More
BIOHIPPO Mouse TMEM119 Microsample
SDP
Catalog No. 5000313969
Encompass
Mouse TMEM119 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMEM119 in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TMEM119 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TMEM119 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TMEM119 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13969 Supplier BIOHIPPO Supplier No. BHE15210659
Mouse TMEM119 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMEM119 in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TMEM119 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TMEM119 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TMEM119 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color