Learn More
BIOHIPPO Mouse TMOD3 Microsample
SDP
Catalog No. 5000315673
Encompass
Mouse TMOD3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMOD3 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 11 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TMOD3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TMOD3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TMOD3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15673 Supplier BIOHIPPO Supplier No. BHE15211056
Mouse TMOD3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMOD3 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 11 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TMOD3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TMOD3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TMOD3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color