Learn More
Sino Biological Mouse TNF ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502372338
Encompass_Preferred
Each vial contains 68 ng of recombinant Mouse TNF-alpha / TNFA / TNFSF2 ELISA Pair Set. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 6000 pg/mL is recommended.
Catalog No. 50-237-2338 Supplier Sino Biological Supplier No. SEK503495
Each vial contains 68 ng of recombinant Mouse TNF-alpha / TNFA / TNFSF2 ELISA Pair Set. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 6000 pg/mL is recommended.