Learn More
Mouse TNF ELISPOT Pair, BD
Catalog No. BDB551875 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 x 96-well Plates
Catalog No. Quantity
BDB551875 5 x 96-well Plates
1 options
Catalog No. BDB551875 Supplier BD Biosciences Supplier No. 551875
Only null left
Add to Cart
Add to Cart
Includes
Mouse TNF ELISPOT Capture Antibody (no azide/low endotoxin), Mouse TNF ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations
Convenient set of unlabelled anti-TNF capture antibody and a biotinylated anti-TNF detection antibody
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 x 96-well Plates