Learn More
Description
The BD CBA Mouse TNF Enhanced Sensitivity Flex Set is a bead-based immunoassay capable of measuring mouse Tumor Necrosis Factor (TNF) in serum and cell culture supernatant samples.
Mouse
Specifications
Specifications
| Antigen | TNF |
| Cross Reactivity | Mouse |
| Includes | 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Measures Mouse tumor necrosis factor, flow cytometry |
| Product Type | Mouse TNF enhanced sensitivity flex set (bead C8) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.