Learn More
Sino Biological Mouse TNF MAb 200ug (Sino Biological)
Supplier:  Sino Biological 50349RN023200
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TNF-alpha/TNFA/TNFSF2 (rM TNF-alpha/TNFA/TNFSF2; Leu80-Leu235; NP_038721.1; Catalog#50349-MNAE).
Catalog No. 50-237-3454