Learn More
Sino Biological Recombinant Anti-TNF-alpha Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50349R02320
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TNF-alpha / TNFA / TNFSF2 (rM TNF-alpha / TNFA / TNFSF2; Catalog#50349-MNAE; NP_038721.1; Leu80-Leu235).
Catalog No. 50-240-8756