Learn More
Sino Biological Mouse TNFR1/CD120a Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785875
Encompass_Preferred
The Mouse TNFRSF1A ELISA Pair Set is for the quantitative determination of Mouse TNFRSF1A.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse TNFRSF1A coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse TNFRSF1A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse TNFRSF1A monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse TNFRSF1A present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85875 Supplier Sino Biological Supplier No. SEK504965
The Mouse TNFRSF1A ELISA Pair Set is for the quantitative determination of Mouse TNFRSF1A.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse TNFRSF1A coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse TNFRSF1A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse TNFRSF1A monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse TNFRSF1A present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.