Learn More
Sino Biological Recombinant Mouse CD55/DAF Protein (His Tag)
Supplier:  Sino Biological 50468M08H1
Encompass_Preferred
A DNA sequence encoding the mature form of mouse CD55 (Q61475) (Met 1-Thr 361) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-222-3730