Learn More
ACROBiosystems Mouse TNFR2 Protein Fc Tag
Supplier:  ACROBiosystems TN2M525450UG
Encompass_Preferred
Mouse TNFR2 Protein Fc Tag (TN2-M5254) is expressed from human 293 cells (HEK293). It contains AA Val 23 - Gly 258 (Accession # P25119-1).
Catalog No. 50-253-4031