Learn More
Sino Biological Anti-DR5/TRAIL R2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50412RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TNFRSF10B (rM TNFRSF10B; Catalog#50412-M08H; NP_064671.2; Met 1-Ser 177). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3279