Learn More
Sino Biological Anti-DR5/TRAIL R2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50412T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse DR5/TNFRSF10B (rM DR5/TNFRSF10B; Catalog#50412-M08H; NP_064671.2; Met1-Ser177). DR5/TNFRSF10B specific IgG was purified by Mouse DR5/TNFRSF10B affinity chromatography.
Catalog No. 50-255-3277