Learn More
Sino Biological Recombinant Mouse HVEM Protein (His & hFc Tag)
Supplier:  Sino Biological 10567M03H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 206) of mouse HVEM (NP_849262.1) precursor was fused with C-terminal His-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-240-8618