Learn More
Sino Biological Recombinant Mouse BCMA Protein (hFc Tag), HPLC-verified 1
Supplier:  Sino Biological 50076M01H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Thr 49) of mouse BCMA (NP_035738.1) precursor was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-224-7207