Learn More
Sino Biological Recombinant Mouse BCMA Protein (ECD, hFc & AVI Tag), Biotinylated
Supplier:  Sino Biological 50076M42HB20
Encompass_Preferred
A DNA sequence encoding the Mouse TNFRSF17 (NP_035738.1) (Met1-Thr49) was expressed with a c-terminal Fc region of human IgG1 tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-3478