Learn More
Sino Biological Anti-TROY/TNFRSF19 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50148RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TNFRSF19 (rM TNFRSF19; Catalog#50148-M08H; NP_001157627.1; Met 1-Leu 170). TNFRSF19 specific IgG was purified by mouse TNFRSF19 affinity chromatography.
Catalog No. 50-255-3404