Learn More
Sino Biological Recombinant Mouse TROY/TNFRSF19 Protein (ECD, mFc Tag)
Supplier:  Sino Biological 50148M38H500
Encompass_Preferred
A DNA sequence encoding the mouse TNFRSF19 (NP_001157627.1) (Met1-Leu170) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-255-3409