Learn More
Sino Biological Recombinant Anti-TNFR1/CD120a Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50496R10420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TNFR1 / TNFRSF1A / CD120a (rM TNFR1 / TNFRSF1A / CD120a; Catalog#50496-M08H; NP_035739.2; Met1-Ala212).
Catalog No. 50-241-2177