Learn More
Sino Biological Recombinant Anti-TNFR-2 / CD120b Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50128R20420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TNFR2 / TNFRSF1B / CD120b (rM TNFR2 / TNFRSF1B / CD120b; Catalog#50128-M08H; NP_035740.2; Met1-Gly258).
Catalog No. 50-241-1695