Learn More
Sino Biological Mouse TNFRSF1B MAb 25TS (Sino Biological)
Supplier:  Sino Biological 202377T08100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TNFR2 / CD120b / TNFRSF1B (rM TNFR2 / CD120b / TNFRSF1B; Catalog#50128-M08H; NP_035740.2; Met1-Gly258) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-4296