Learn More
Sino Biological Mouse TNFRSF1B PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50128RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TNFR2 / CD120b / TNFRSF1B (rM TNFR2 / CD120b / TNFRSF1B; Catalog#50128-M08H; NP_035740.2; Met 1-Gly 258). TNFR2 / CD120b / TNFRSF1B specific IgG was purified by Mouse TNFR2 / CD120b / TNFRSF1B affinity chromatography.
Catalog No. 50-237-3096