Learn More
Sino Biological Mouse TNFRSF4 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 50808R055F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse OX40 (rM OX40; Catalog#50808-MCCH; NP_035789.1; Met1-Pro211) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-237-2829