Learn More
Sino Biological Mouse TNFRSF9 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 5A7656M49HB100
Encompass_Preferred
A DNA sequence encoding the Mouse TNFRSF9 (NP_001070977.1) (Met1-Leu187) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-2510