Learn More
Sino Biological Recombinant Mouse 4-1BB/CD137 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 50811M49HB20
Encompass_Preferred
A DNA sequence encoding the Mouse TNFRSF9 (NP_001070976.1)(Met1-Leu211) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-0054