Learn More
Sino Biological Anti-TRAIL/TNFSF10 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50166T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TNFSF10 / TRAIL / APO-2L (CD253) (Catalog#50166-M07E; P50592; Pro118-Asn291). TNFSF10 / TRAIL / APO-2L (CD253) specific IgG was purified by Mouse TNFSF10 / TRAIL / APO-2L (CD253) affinity chromatography.
Catalog No. 50-241-1655