Learn More
Sino Biological Recombinant Mouse APRIL/TNFSF13 Protein (hFc Tag)
Supplier:  Sino Biological 50965M01H1
Encompass_Preferred
A DNA sequence encoding the mouse TNFSF13 (Q5F2A4) (Ala96-Leu240) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-255-3043