Learn More
Sino Biological Mouse TNFSF9 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 50067R01420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TNFSF9 / 4-1BBL (rM TNFSF9; Catalog#50067-M07H; Arg 104-Glu 309; NP_033430.1).
Catalog No. 50-237-2388