Learn More
Sino Biological Anti-4-1BB Ligand/TNFSF9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50067RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TNFSF9 / 4-1BBL extracellular domain (rM TNFSF9; Catalog#50067-M07H; Arg 104-Glu 309; NP_033430.1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0107