Learn More
Biomatik Corporation Mouse Toll Like Receptor 1 (TLR1) ELISA Kit
Supplier:  Biomatik Corporation EKU07745
Encompass_Preferred
Mouse Toll Like Receptor 1 (TLR1) ELISA Kit for detecting TLR1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EPQ1. Gene ID: 21897. Target Synonyms: CD281; TIL; Rsc786; Toll/interleukin-1 receptor-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5897