Learn More
Biomatik Corporation Mouse Toll Like Receptor 9 (TLR9) ELISA Kit
Supplier:  Biomatik Corporation EKU09019
Encompass_Preferred
Mouse Toll Like Receptor 9 (TLR9) ELISA Kit for detecting TLR9 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EQU3. Gene ID: 81897. Target Synonyms: CD289. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8576